LIFH0C9551F0 Life Chemicals Inc.

2,3,4-trimethyl-1,5-naphthyridine

  • MDL number MFCD26381830
  • Cas Number : 1557262-60-8
  • Molecular Weight : 172.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart