LIFH0C95520A Life Chemicals Inc.

3-(1-phenyl-1H-pyrazol-4-yl)propan-1-amine

  • MDL number MFCD08572192
  • Cas Number : 400866-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart