LIFH11AADBC0 Life Chemicals Inc.

5-(bromomethyl)-2,3-dihydro-1,4-benzodioxine

  • MDL number MFCD05664400
  • Cas Number : 214894-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart