LIFH11AADBD5 Life Chemicals Inc.

5-bromo-4-methoxypyridine-2-carboxylic acid

  • MDL number MFCD18257512
  • Cas Number : 1211586-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart