LIFH11AADBDC Life Chemicals Inc.

2-sulfanyl-1H-1,3-benzodiazol-6-ol

  • MDL number MFCD04117895
  • Cas Number : 92806-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 91
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart