LIFH11AADBE1 Life Chemicals Inc.

5-bromo-3-chloro-2-fluoropyridine-4-carbaldehyde

  • MDL number MFCD29059499
  • Cas Number : 2092512-51-9
  • Molecular Weight : 238.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart