LIFH11E4EF0F Life Chemicals Inc.

tert-butyl 4-(3-cyanopyridin-2-yl)piperidine-1-carboxylate

  • MDL number MFCD31660512
  • Cas Number : 2303565-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart