LIFH11E4EF17 Life Chemicals Inc.

1-(4-methoxyphenyl)piperazin-2-one hydrochloride

  • MDL number MFCD29052278
  • Cas Number : 1284247-69-3
  • Molecular Weight : 242.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart