LIFH11E4EF2D Life Chemicals Inc.

2,5-dimethyl-2H,4H,5H,6H,7H-pyrazolo[3,4-d]pyrimidine-4,6-dione

  • MDL number MFCD24549973
  • Cas Number : 36075-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart