LIFH11E4EF42 Life Chemicals Inc.

2-hydroxybenzene-1,3-dicarbonitrile

  • MDL number MFCD16999196
  • Cas Number : 28177-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart