LIFH22CC184C Life Chemicals Inc.

4-(5-amino-2-methylphenoxy)benzoic acid

  • Cas Number : 2303565-76-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart