LIFH22CC1855 Life Chemicals Inc.

1-(7-bromo-2,3-dihydro-1-benzofuran-2-yl)methanamine hydrochloride

  • Cas Number : 2303565-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart