LIFH22CC1857 Life Chemicals Inc.

3-bromo-5-fluoro-[1,1'-biphenyl]-4-carbaldehyde

  • MDL number MFCD31696197
  • Cas Number : 2244721-36-4
  • Molecular Weight : 279.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart