LIFH22CC1858 Life Chemicals Inc.

tert-butyl 4-(3-cyanophenyl)-3-methylpiperazine-1-carboxylate

  • Cas Number : 2303565-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart