LIFH22CC185D Life Chemicals Inc.

(6-phenylpyridin-3-yl)methanamine

  • MDL number MFCD09025823
  • Cas Number : 294648-38-7
  • Molecular Weight : 184.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart