LIFH22CC1863 Life Chemicals Inc.

(4-fluorooxan-4-yl)methanol

  • MDL number MFCD18072006
  • Cas Number : 883442-46-4
  • Molecular Weight : 134.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart