LIFH22CC1873 Life Chemicals Inc.

1-(tetrahydro-2H-thiopyran-4-yl)-1,4-diazepane dihydrochloride

  • MDL number MFCD29046663
  • Cas Number : 2098093-19-5
  • Molecular Weight : 273.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart