LIFH22CC188E Life Chemicals Inc.

ethyl 3,3-difluorocyclobutane-1-carboxylate

  • MDL number MFCD16660028
  • Cas Number : 681128-38-1
  • Molecular Weight : 164.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart