LIFH22CC1897 Life Chemicals Inc.

6-(trifluoromethyl)-1',2',3',6'-tetrahydro-2,4'-bipyridine hydrochloride

  • MDL number MFCD31660519
  • Cas Number : 2303565-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart