LIFH22CC1899 Life Chemicals Inc.

1,3-dimethyl 2-(3-oxocyclopentyl)propanedioate

  • MDL number MFCD00482100
  • Cas Number : 111209-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart