LIFH22CC189B Life Chemicals Inc.

2-(2-ethyl-4,5,6,7-tetrahydro-1H-1,3-benzodiazol-1-yl)acetic acid hydrochloride

  • Cas Number : 2303565-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart