LIFH22CC189F Life Chemicals Inc.

5-methyl-2-azaspiro[3.3]heptane; oxalic acid

  • Cas Number : 2303565-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart