LIFH22CC18AA Life Chemicals Inc.

ethyl 4-chloro-2-methoxypyrimidine-5-carboxylate

  • MDL number MFCD12923009
  • Cas Number : 65269-18-3
  • Molecular Weight : 216.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart