LIFH22CC18B9 Life Chemicals Inc.

2,4-difluoro-[1,1'-biphenyl]-3-carbaldehyde

  • MDL number MFCD31619493
  • Cas Number : 2221812-11-7
  • Molecular Weight : 218.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart