LIFH22CC18C2 Life Chemicals Inc.

4-(4-methoxyphenyl)piperidine

  • MDL number MFCD03839927
  • Cas Number : 67259-62-5
  • Molecular Weight : 191.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart