LIFH22CC18C6 Life Chemicals Inc.

2-amino-1-(3-nitrophenyl)ethan-1-one hydrochloride

  • MDL number MFCD00051997
  • Cas Number : 36765-84-1
  • Molecular Weight : 216.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart