LIFH22CC18C7 Life Chemicals Inc.

N-(cyclopropylmethyl)-4-methylpyridin-2-amine

  • MDL number MFCD15525724
  • Cas Number : 1250110-40-7
  • Molecular Weight : 162.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart