LIFH22CC18CE Life Chemicals Inc.

(7-bromo-5-fluoro-2,3-dihydro-1-benzofuran-2-yl)methanol

  • MDL number MFCD27929938
  • Cas Number : 852111-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart