LIFH22CC18D8 Life Chemicals Inc.

N,N,1-trimethyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine

  • MDL number MFCD05938808
  • Cas Number : 90085-12-4
  • Molecular Weight : 177.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart