LIFH2F69E1B0 Life Chemicals Inc.

ethyl 2-(azetidin-3-yl)-1,3-thiazole-4-carboxylate; trifluoroacetic acid

  • Cas Number : 2303565-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart