LIFH2F69E1B1 Life Chemicals Inc.

1-[5-(aminomethyl)naphthalen-1-yl]methanamine dihydrochloride

  • Cas Number : 108629-56-7
  • Molecular Weight : 259.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart