LIFH2F69E1C0 Life Chemicals Inc.

trans-4-aminooxolan-3-ol

  • MDL number MFCD09260577
  • Cas Number : 153610-14-1
  • Molecular Weight : 103.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart