LIFH2F69E1D6 Life Chemicals Inc.

(cyclobutylmethyl)(methyl)amine hydrochloride

  • MDL number MFCD16817426
  • Cas Number : 1251925-47-9
  • Molecular Weight : 135.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart