LIFH2F69E1EA Life Chemicals Inc.

4H,5H,6H,7H-pyrazolo[1,5-a]pyridine-3-carboxamide

  • MDL number MFCD31672330
  • Cas Number : 1866182-74-2
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart