LIFH2F69E200 Life Chemicals Inc.

[2-(hydroxymethyl)spiro[3.3]heptan-2-yl]methanol

  • Cas Number : 93225-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart