LIFH2F69E204 Life Chemicals Inc.

3-methyl-2-oxopyrrolidine-3-carbohydrazide

  • Cas Number : 2303565-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart