LIFH2F69E208 Life Chemicals Inc.

5-(2,4-dichlorophenyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD00475836
  • Cas Number : 28004-63-9
  • Molecular Weight : 246.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart