LIFH2F69E209 Life Chemicals Inc.

(1R)-N-[(1R)-1-(4-methoxyphenyl)ethyl]-8-azaspiro[4.5]decan-1-amine

  • MDL number MFCD30472136
  • Cas Number : 2007920-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart