LIFH2F69E212 Life Chemicals Inc.

2-chloro-N-cyclohexylacetamide

  • MDL number MFCD00085678
  • Cas Number : 23605-23-4
  • Molecular Weight : 175.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart