LIFH2F69E214 Life Chemicals Inc.

2,6-difluoro-3-formylbenzonitrile

  • MDL number MFCD26687933
  • Cas Number : 1541976-46-8
  • Molecular Weight : 167.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart