LIFH2F69E21A Life Chemicals Inc.

5-(3-bromophenyl)-1,2,4-thiadiazole

  • Cas Number : 101494-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart