LIFH2F69E220 Life Chemicals Inc.

2-(2-methoxyphenyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD11895894
  • Cas Number : 92016-93-8
  • Molecular Weight : 192.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart