LIFH2F69E228 Life Chemicals Inc.

5-[(4-fluorophenyl)methyl]-1,2-oxazole-3-carboxylic acid

  • Cas Number : 1621013-99-7
  • Molecular Weight : 221.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart