LIFH3031A5BA Life Chemicals Inc.

2-chloro-5-(cyanomethyl)benzoic acid

  • MDL number MFCD20484285
  • Cas Number : 66658-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart