LIFH3031A5C4 Life Chemicals Inc.

7-bromo-1H,2H,3H-pyrrolo[3,2-c]pyridine

  • MDL number MFCD20687373
  • Cas Number : 1198293-24-1
  • Molecular Weight : 199.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart