LIFH3035C0EA Life Chemicals Inc.

2-(3,3-difluorocyclobutyl)acetonitrile

  • MDL number MFCD24856416
  • Cas Number : 1219019-02-9
  • Molecular Weight : 131.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart