LIFH3035C0F5 Life Chemicals Inc.

4-(dimethylamino)cyclohexan-1-ol

  • MDL number MFCD09037891
  • Cas Number : 61168-09-0
  • Molecular Weight : 143.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart