LIFH3035C111 Life Chemicals Inc.

4-bromo-2,3,5,6-tetrafluorobenzaldehyde

  • MDL number MFCD31652790
  • Cas Number : 108574-98-7
  • Molecular Weight : 256.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart