LIFH309942B0 Life Chemicals Inc.

4-aminopyridine-2-carboxylic acid

  • MDL number MFCD02685671
  • Cas Number : 100047-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart