LIFH309942B4 Life Chemicals Inc.

trans-2-(phenylamino)cyclopentan-1-ol

  • MDL number MFCD20661803
  • Cas Number : 210408-12-1
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart